Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With DIN Thread
Click to view available options
Diameter (Metric) Outer:
64 mm
85 mm
105 mm
Unit Size:
Pack of 10
Description
- With DIN thread (GL) and screw cap (red)
- With scale (approx. volume /ml)
- With inscription label
- Material: DURAN™ (flask), PBT (cap)
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Thread Size | 32 GL |
| Diameter (Metric) Outer | 85 mm |
| Quantity | 10/Pk. |