Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With Conical Ground Joint
Product Code. 10652662
Change view
Click to view available options
Capacity (Metric):
150 mL
200 mL
2 L
Standard Taper Joint:
NS 19/26
NS 24/29
Socket NS 19/26
Socket NS 24/29
Socket NS 29/32
Unit Size:
Pack of 10
Product Code. Capacity (Metric) Standard Taper Joint unitSize
10652662 2 L NS 24/29 Pack of 10
10758081 200 mL Socket NS 19/26 Pack of 10
11933371 150 mL NS 19/26 Pack of 10
11943371 150 mL Socket NS 24/29 Pack of 10
11953371 150 mL Socket NS 29/32 Pack of 10
5 options
Product Code. 10652662
Brand: Lenz Laborglasintrumente 03022473
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN EN ISO 4797 and in addition to standard
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™
Specifications
Capacity (Metric) 2 L
Includes Conical ground joint
No. per Pack 10/Pk.
Standard Taper Joint NS 24/29
Diameter (Metric) Outer 166 mm