Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks, Narrow Neck
Product Code. 10737711
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
200 mL
300 mL
500 mL
1 L
3 L
Height (Metric):
75 mm
90 mm
105 mm
131 mm
156 mm
180 mm
220 mm
310 mm
Unit Size:
Pack of 10
Pack of 2
Product Code. 10737711
Brand: Lenz Laborglasintrumente 07020028
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 1773
  • With beaded rim
  • With graduation
  • Material: DURAN™
Specifications
Diameter (Metric) Outer Neck 22 mm
Capacity (Metric) 50 mL
Material Duran Glass
Height (Metric) 90 mm
Quantity 10/Pk.