Learn More
Reitenspiess™ Erlenmeyer Flasks and Bottles Brush
Product Code. 10271221
Change view
Click to view available options
Length (Metric):
470 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10271221 470 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10271221 Supplier Reitenspiess™ Supplier No. 23630234
  • ONLINE EXCLUSIVE PRICE
    244.14 NOK
    99.25 NOK / Pack of 1
    Save 144.89 NOK
    59% Off
Estimated Shipment: 16-11-2026
Reitenspiess™ Erlenmeyer Flasks and Bottles Brush – has a 60mm head size and is a useful tool in the laboratory for cleaning, disinfecting and sterilizing Ehrlenmeyer laboratory glassware.
  • 60mm diameter
  • For Eherlenmeyer flask cleaning, disinfecting, sterilizing
  • Sterile
Specifications
Product Type Erlenmeyer Flasks and Bottles
Diameter (Metric) Bristle 60 mm
For Use With (Application) Cleaning, disinfection, sterilization
Length (Metric) 470 mm
Quantity 1 Ea.