Learn More
OHAUS™ Erlenmeyer Flask Clamp
Product Code. 15772509 Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
1 L Erlenmeyer Flask
2 L Erlenmeyer Flask
2.8 L Erlenmeyer Flask
3 L Erlenmeyer Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
6 L Erlenmeyer Flask
Unit Size:
Each
Product Code. 15772509
Brand: OHAUS™ 30400095
Only null left
Add to basket
Add to basket
OHAUS™ Erlenmeyer Flask Clamp is designed to securely grip and position various sizes of flasks.
  • Constructed of stainless steel
  • For use with flasks
  • Designed to securely hold flask in place
TRUSTED_SUSTAINABILITY
Specifications
Type Flask Clamp
Material Stainless Steel
For Use With (Equipment) 5 L Erlenmeyer Flask
For Use With (Application) Holding flasks
Description 5 Liter Erlenmeyer Flask Clamp
Color Silver
Jaw Type Four Interlocking