Description
ELAC2 Polyclonal specifically detects ELAC2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunoprecipitation/Western Blot.
Specifications
Specifications
| Antigen | ELAC2 |
| Applications | Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ELAC2 fragment. |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 60528 |
| Target Species | Human |
| Content And Storage | Store at 4C in the dark. |
| Product Type | Antibody |
| Isotype | IgG |
| Product Line | Novus |