missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERGIC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16029-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 50 kDa, and this antibody specifically recognizes the 50 kDa protein.
ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.Specifications
| ERGIC3 | |
| Polyclonal | |
| Unconjugated | |
| ERGIC3 | |
| C20orf47, CGI 54, ERGIC and Golgi 3, ERGIC3, Erv46, NY BR 84, PRO0989, SDBCAG84 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 296306, 51614, 66366 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CQE7, Q9Y282 | |
| ERGIC3 | |
| ERGIC3 Fusion Protein Ag8893 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction