missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Origene Technologies ERAP1 Monoclonal Antibody (OTI6E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812889S
Additional Details : Weight : 0.05000kg
Description
ERAP1 Monoclonal antibody specifically detects ERAP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ERAP1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9EQH2, Q9JJ22, Q9NZ08 | |
Mouse | |
Protein A/G | |
RUO | |
51752, 80897, 80898 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI6E3 | |
Unconjugated | |
Erap1 | |
adipocyte-derived leucine aminopeptidase; ALAP; A-LAP; aminopeptidase PILS; aminopeptidase regulator of TNFR1 shedding; Appils; Arts1; ARTS-1; CTD-2260A17.2; endoplasmic reticulum aminopeptidase 1; endoplasmic reticulum aminopeptidase 1 delta-Exon-11 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-13 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-14 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-15 isoform; endoplasmic reticulum aminopeptidase associated with antigen processing; ERAAP; ERAAP1; ER-aminopeptidase 1; ERAP1; KIAA0525; leucyl-specific aminopeptidase PILS; PILSA; PILSAP; PILS-AP; Puromycin-insensitive leucyl-specific aminopeptidase; type 1 tumor necrosis factor receptor shedding aminopeptidase regulator; UNQ584/PRO1154; VEGF-induced aminopeptidase | |
Human recombinant protein fragment corresponding to amino acids 529-941 of human ERAP1 (NP_001035548) produced in E.coli. | |
30 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |