Learn More
Thermo Scientific™ Crimper and Decrimper Jaw Sets for Programmable Electronic High-Power Crimp Stations
Product Code. 15227308
Change view
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
Type:
Electronic Crimper
Electronic Crimper (Flip-off)
Electronic Decrimper
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Type Unit Size
15227308 11 mm Electronic Decrimper Each
15207318 8 mm Electronic Crimper Each
15257308 11 mm Electronic Crimper Each
15267308 13 mm Electronic Crimper Each
15237308 13 mm Electronic Decrimper Each
15277308 13 mm Electronic Crimper (Flip-off) Each
15287308 20 mm Electronic Crimper Each
15247308 20 mm Electronic Decrimper Each
15297308 20 mm Electronic Crimper (Flip-off) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15227308 Supplier Thermo Scientific™ Supplier No. ECDJ11
Estimated Shipment: 27-11-2026
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Stations with these crimper and decrimper sets.
Optimize your Thermo Scientific™ Programmable Electronic High-Power Crimp Station with these crimper and decrimper sets.
Available in a variety of sizes and formats.
TRUSTED_SUSTAINABILITY
Specifications
No. per Pack 1 Ea.
Type Electronic Decrimper
For Use With (Equipment) Programmable Electronic High-Power Crimp Station, 11 mm Vial Closure
Size 11 mm
For Use With (Application) Industrial Chromatography