Learn More
EPS8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12455-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.Specifications
EPS8 | |
Polyclonal | |
Unconjugated | |
EPS8 | |
EPS8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2059 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.49 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12929 | |
EPS8 | |
EPS8 Fusion Protein Ag3121 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |