missing translation for 'onlineSavingsMsg'
Learn More

EPS8 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12455-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890174

  • 1281.92 NOK / 20µL
Estimated Shipment: 13-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
Specifications

Specifications

EPS8
Polyclonal
Unconjugated
EPS8
EPS8
Rabbit
Antigen Affinity Chromatography
RUO
2059
-20°C
Liquid
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12929
EPS8
EPS8 Fusion Protein Ag3121
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers