Learn More
Epoxyazadiradione, TRC
Product Code. 30136049
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136049 5 mg 5mg
30136925 10 mg 10mg
30007071 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136049 Supplier Toronto Research Chemicals Supplier No. TRCE5900185MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18385-59-6
Molecular Formula C28 H34 O6
Molecular Weight (g/mol) 466.56
Synonym Nimbinin, 14,15-Epoxy-14H-cyclopenta[a]phenanthrene, 24-norchola-1,20,22-triene-3,16-dione deriv., Azadiradione 14β,15β-Epoxide
SMILES CC(=O)O[C@@H]1C[C@H]2C(C)(C)C(=O)C=C[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](C(=O)[C@H]5O[C@@]45[C@]13C)c6cocc6
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C28H34O6/c1-15(29)33-20-13-18-24(2,3)19(30)8-10-25(18,4)17-7-11-26(5)21(16-9-12-32-14-16)22(31)23-28(26,34-23)27(17,20)6/h8-10,12,14,17-18,20-21,23H,7,11,13H2,1-6H3/t17-,18+,20-,21-,23-,25-,26+,27+,28-/m1/s1
Formula Weight 466.2355
Chemical Name or Material Epoxyazadiradione