Learn More
Epredia™ Epoxy Diagnostic Slides
Product Code. 13535770 Shop All Epredia Products
Click to view available options
Well Count:
1
3
4
5
8
10
12
Color:
Black
Blue
White
Diameter (Metric):
10 mm Well
11.43 mm Well
13 mm Well
14 mm Well
5 mm Well
6 mm Well
6.7 mm Well
8 mm Well
Unit Size:
Pack of 100
Pack of 1000
Product Code. 13535770
Brand: Epredia™ X1XES333W
Only null left
Add to basket
Add to basket
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use
  • Binds and keeps liquids applied to the well area
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 25 mm
Width (Metric) 75 mm
Material Epoxy
Thickness (Metric) 1 mm
Type Diagnostic Slide
Well Count 1
Color White
Diameter (Metric) 13 mm Well