Description
- Manufactured from epoxy-coated steel wire
- Rubber feet prevent slippage.
- Available with polypropylene or polyamide bags
Specifications
Specifications
| Description | Epoxy coated |
| Autoclavable | Not Autoclavable |
| Color | White |
| For Use With (Equipment) | Disposal bags |
| Includes | 100 polypropylene disposal bags |
| Material | Steel Wire |
| Height (Metric) | 250 mm |
| Diameter (Metric) | 12 cm |
| Product Type | Stand |