Learn More
Epinephrine Sulfonic Acid, TRC
Product Code. 17971235
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971235 25 mg 25mg
17982085 100 mg 100mg
17900835 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17971235 Supplier Toronto Research Chemicals Supplier No. TRCE58860525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26405-77-6
Molecular Formula C9 H13 N O5 S
Molecular Weight (g/mol) 247.27
Synonym Benzenemethanesulfonic acid, 3,4-dihydroxy-α-[(methylamino)methyl]-, α-Toluenesulfonic acid, 3,4-dihydroxy-α-[(methylamino)methyl]- (6CI,8CI), 3,4-Dihydroxy-α-[(methylamino)methyl]benzenemethanesulfonic acid, 1-(3,4-Dihydroxy-phenyl)-2-methylamino-ethanesulfonic acid, 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethane-1-sulfonic acid, 1-(3,4-Dihydroxyphenyl)-2-(methylamino)ethanesulfonic acid, Epinephrine sulfonic acid, Adrenaline sulfonic acid
IUPAC Name 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethanesulfonic acid
SMILES CNCC(c1ccc(O)c(O)c1)S(=O)(=O)O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C9H13NO5S/c1-10-5-9(16(13,14)15)6-2-3-7(11)8(12)4-6/h2-4,9-12H,5H2,1H3,(H,13,14,15)
Formula Weight 247.0514
Percent Purity >95%
Chemical Name or Material Epinephrine Sulfonic Acid
Grade HPLC