missing translation for 'onlineSavingsMsg'
Learn More

Epetraborole hydrochloride, MedChemExpress

Product Code. p-200099602
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497917

Brand: Medchem Express HY12479A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Epetraborole hydrochloride is a novel leucyl-tRNA synthetase (LeuRS) inhibitor, which inhibits protein synthesis by binding to the terminal adenosine ribose (A76) of leucyl-tRNA synthetase.

Epetraborole hydrochloride is a novel leucyl-tRNA synthetase (LeuRS) inhibitor, which inhibits protein synthesis by binding to the terminal adenosine ribose (A76) of leucyl-tRNA synthetase.

Specifications

Chemical Name or Material Epetraborole hydrochloride
CAS 1234563-16-6
Color White
Purity Grade Notes Research
Molecular Formula C11H17BClNO4
Quantity 5 mg
Synonym GSK2251052 hydrochloride
Solubility Information DMSO : 200 mg/mL (731.21 mM; Need ultrasonic) ∣H2O : ≥ 28 mg/mL (102.37 mM)
SMILES OCCCOC1=C(B(O)O[C@@H]2CN)C2=CC=C1.[H]Cl
Molecular Weight (g/mol) 273.52
Formula Weight 273.52
Percent Purity 99.25%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.