Learn More
Epertinib, MedChemExpress
Product Code. 16441079
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
16441079 1 mg 1mg
1 options
Product Code. 16441079
Brand: Medchem Express HY107367
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Epertinib (S-22611) is a potent, oral, reversible, and selective tyrosine kinase inhibitor of EGFR, HER2 and HER4, with IC50s of 1.48 nM, 7.15 nM and 2.49 nM, respectively. Epertinib shows potent antitumor activity.
Epertinib (S-22611) is a potent, oral, reversible, and selective tyrosine kinase inhibitor of EGFR, HER2 and HER4, with IC50s of 1.48 nM, 7.15 nM and 2.49 nM, respectively. Epertinib shows potent antitumor activity.
Specifications
Chemical Name or Material Epertinib
CAS 908305-13-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H27ClFN5O3
Quantity 1 mg
Synonym S-22611
SMILES CC#C/C(C1=CC2=C(NC3=CC=C(OCC4=CC=CC(F)=C4)C(Cl)=C3)N=CN=C2C=C1)=N\OC[C@@H]5NCCOC5
Molecular Weight (g/mol) 560.02
Formula Weight 560.02
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease