Learn More
Eperezolid, MedChemExpress
Product Code. 16424946
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16424946
Brand: Medchem Express HY10393
MedChemExpress Eperezolid(PNU-100592) is a oxazolidinone antibacterial agent, Eperezolid demonstrated good in vitro inhibitory activity, regardless of methicillin susceptibility for staphylococci(MIC90= 1-4 mg/ml).
Eperezolid(PNU-100592) is a oxazolidinone antibacterial agent, Eperezolid demonstrated good in vitro inhibitory activity, regardless of methicillin susceptibility for staphylococci(MIC90= 1-4 mg/ml).
Specifications
Chemical Name or Material Eperezolid
Physical Form Solid
CAS 165800-04-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H23FN4O5
For Use With (Application) COVID-19-immunoregulation
Synonym PNU-100592
Solubility Information DMSO : ≥ 44 mg/mL (111.56 mM)
SMILES O=C(O[C@H]1CNC(C)=O)N(C1)C2=CC(F)=C(C=C2)N3CCN(CC3)C(CO)=O
Molecular Weight (g/mol) 394.4
Formula Weight 394.4
Percent Purity 95.77%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.