Description
- Rubber (EPDM) construction
- Conical shape
- Provides a secure fit between filter funnel and filter flask
- Supplied in packs of 10
Specifications
Specifications
| For Use With (Application) | Used for Filter Funnels and Filter Flasks |
| Length (Metric) | 28 mm |
| Quantity | 10 |
| Diameter (Metric) Outside | 16 mm |
| Material | EPDM |
| Type | Gasket, Conical |