Learn More
BRAND™ EPDM Rubber Conical Gaskets
Product Code. 10277971 Shop All Brand Products
Change view
Click to view available options
Diameter (Metric) Outside:
11 mm
16 mm
20 mm
22 to 84 mm
25 mm
32 mm
43 mm
71 mm
89 mm
Unit Size:
Pack of 10
Pack of 8
Product Code. Diameter (Metric) Outside unitSize
10277971 71 mm Pack of 10
10548781 11 mm Pack of 10
10689071 16 mm Pack of 10
10518971 20 mm Pack of 10
10325571 43 mm Pack of 10
10101971 25 mm Pack of 10
10049900 32 mm Pack of 10
10614552 22 to 84 mm Pack of 8
10599531 89 mm Pack of 10
9 options
Product Code. 10277971
Brand: BRAND™ 463139
Only null left
Add to basket
Add to basket
Place between filter funnel and filter flask for a secure fit.
  • Rubber (EPDM) construction
  • Conical shape
  • Provides a secure fit between filter funnel and filter flask
  • Supplied in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inside 71 x 52 mm dia.
For Use With (Application) Used for Filter Funnels and Filter Flasks
Quantity 10
Size (Metric) 71 mm
Diameter (Metric) Outside 71 mm
Material EPDM
Type Gasket, Conical