Specifications
| Type | End Piece Complete |
| Packaging Quantity | 1 Each |
| Includes | 1 x Locking ring, 1 x washer, 1 x sealing ring, 1 x end piece, 1 x O-ring 25.07 x 2.62 mm |
| For Use With | XK16 Columns |
| Material | EPDM, PTFE, POM |