Learn More
EPA Method 515.4 Herbicide Mixture 408 10-100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4,5-Trichlorophenoxyacetic acid∣2,4-Dichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣Acifluorfen∣Bentazone∣Chloramben∣Chlorthal-diacid∣Dacthal monoacid∣Dalapon∣Dicamba∣Dichlorprop∣Dinoseb∣Fenoprop∣Pentachlorophenol∣Picloram∣Quinchlorac |
| CAS | 93-76-5, 94-75-7, 51-36-5, 94-82-6, 50594-66-6, 25057-89-0 |
| Analyte or Component Names | 2,4,5-Trichlorophenoxyacetic acid∣2,4-Dichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣Acifluorfen∣Bentazone∣Chloramben∣Chlorthal-diacid∣Dacthal monoacid∣Dalapon∣Dicamba∣Dichlorprop∣Dinoseb∣Fenoprop∣Pentachlorophenol∣Picloram∣Quinchlorac |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Pentachlorophenol 10 μg/mL∣2,4,5-Trichlorophenoxyacetic acid 25 μg/mL∣Fenoprop 25 μg/mL∣3,5-Dichlorobenzoic acid 50 μg/mL∣Acifluorfen 50 μg/mL∣Chloramben 50 μg/mL∣Chlorthal-diacid 50 μg/mL∣Dacthal monoacid 50 μg/mL∣Dicamba 50 μg/mL∣Picloram 50 μg/mL∣Quinchlorac 50 μg/mL∣2,4-Dichlorophenoxyacetic acid 100 μg/mL∣4-(2,4-Dichlorophenoxy)butyric acid 100 μg/mL∣Bentazone 100 μg/mL∣Dalapon 100 μg/mL∣Dichlorprop 100 μg/mL∣Dinoseb 100 μg/mL |