Learn More
Eosin Y Disodium Salt, 10% Solution, MP Biomedicals™
Shop All MP Biomedicals ProductsDescription
At concentrations higher than 10-50μM (near 100μM) the affinity for Mg2+-ATPase for the ion-activator is reduced. This compound works as a molecular photoelectrode via catalyzation of the visible-light-driven electron-transfer reaction. This agent also enhances fluorescent quenching abilities by forming a complex with polyvinulpyrrolidone. It is used as a counterstain for hematoxylin.
Eosin Y disodium salt is used in combination with Azure II: to show morphological details of marrow cells; buffered Azure A/Eosin Y method for demostrating cell granules, microorganisms and nuclei; for showing insect musculature; for demonstrating cells infected with mosquito-iridescent virus; use in combination with Alum Hematoxylin/Azure II to show mast cells, early necrobiotic changes in cells and protozoans, rickettsias and bacteria in tissues; to demonstrate volutin granules in any microorganism; Bond's modification of Mann's Eosin-Methyl Blue for Negri bodies in smears; Harris stain for Negri bodies in smears; Mann's method for staining Negri bodies in sections; Gridley's stain for Endamoeba in tissue sections; a modified methenamine-silver nitrate method for the demonstration of Endamoeba in sections.
Specifications
Specifications
| Formula Weight | 691.86 |
| Quantity | 500 mL |
| Physical Form | Solution |
Safety and Handling
- Eosin Y Disodium Salt, 10% Solution
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- MIXTURE LIST-Contains : Eosin Y Disodium Salt
Your input is important to us. Please complete this form to provide feedback related to the content on this product.