Learn More
Invitrogen™ EOMES Monoclonal Antibody (WD1928), Alexa Fluor™ 660, eBioscience™, Invitrogen™

Description
The WD1928 antibody recognizes Eomesodermin (Eomes), also known as T-box brain 2 (TBR2). Eomes is a T-box transcription factor that is highly homologous to T-bet, which is essential during trophoblast development and gastrulation in most vertebrates. In the immune system, Eomes controls the differentiation of effector and memory CD8+ T cells, as well as natural killer (NK) cells. Expression of Eomes in these cells correlates with high expression of CD122, the common beta-chain of the IL-2R and IL-15R.
Specifications
Specifications
| Antigen | EOMES |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | WD1928 |
| Concentration | 5 μL/Test |
| Conjugate | Alexa Fluor 660 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Eomes |
| Gene Accession No. | O95936 |
| Gene Alias | C77258; EOMES; eomesodermin; eomesodermin homolog; eomesodermin homolog (Xenopus laevis); T-box brain protein 2; T-box transcription factor; TBR2; TBR-2; T-brain-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.