missing translation for 'onlineSavingsMsg'
Learn More

EOAI3402143, MedChemExpress

Product Code. p-200096957
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16495326

Brand: Medchem Express HY111408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress EOAI3402143 is a deubiquitinase (DUB) inhibitor, which inhibits dose-dependently inhibits Usp9x/Usp24 and Usp5.

EOAI3402143 is a deubiquitinase (DUB) inhibitor, which inhibits dose-dependently inhibits Usp9x/Usp24 and Usp5.

Specifications

Chemical Name or Material EOAI3402143
CAS 1699750-95-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H28Cl2N4O3
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (99.32 mM; Need ultrasonic)
SMILES ClC1=C(/C=C(C(NC(CCC)C2=CC=C(OCCN3CCOCC3)C=C2)=O)\C#N)N=C(Cl)C=C1.[E]
Molecular Weight (g/mol) 503.42
Formula Weight 503.42
Percent Purity 99.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.