Learn More
Enzyme storage buffer in PBS and glycerol, autoclaved, Thermo Scientific™
Product Code. 15470287
Change view
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15470287 125 mL 125mL
15480287 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15470287 Supplier Thermo Scientific Alfa Aesar Supplier No. J63115.=E
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For frozen storage of enzymes
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Enzyme storage buffer in PBS and glycerol
Physical Form Liquid
Quantity 125 mL