Learn More
Environmental Cal Standard A (Primary): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, U, V, Zn @ 10; Ca, Fe, K, Mg, Na @ 1000 μg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid, VHG™
Product Code. p-200132021
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30003015
Brand: VHG Labs VHGLCAL1A500
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Environmental Cal Standard A (Primary): Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, U, V, Zn @ 10; Ca, Fe, K, Mg, Na @ 1000 μg/mL in 5% HNO3, tr. HF, tr. Tartaric Acid
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL