Learn More
3M™ Enviro Swab ENVSWB100
A pre-moistened swab, ready to use for collecting environmental samples on all types of food contact surfaces in the food industry.
Marque: 3M™ 7100130395
4055.74 NOK valid until 2024-05-31
Use promo code "21654" to get your promotional price.
Informations supplémentaires : Weight : 0.32400kg
Description
- Ready-to-use swab reaches into corners and hard-to-reach areas for increased accuracy.
- Tamper-evident band indicates if the swab has been used.
- Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
- Resealable tube provides tight closure for sample transportation.
- Self-contained swab and vial for increased productivity and efficiency.
Spécification
Swab | |
100/Cs. |