Learn More
Memmert™ Entry Port
Material: Silicone; For Use With: ICO240
Brand:  Memmert™ ICO240F7
Product Code. 15662224
  • ONLINE EXCLUSIVE PRICE
    5460.00 NOK
    4960.28 NOK / Each
    Save 499.72 NOK
    9% Off
Only null left
Estimated Shipment: 09-03-2026
Description
  • Moisture tight
  • Can be closed by silicone stopper
Specifications
Specifications
Silicone
40 mm