Learn More
Sartorius Entris™ II Precision Balance, Internal Adjustment
Shop All Sartorius Products
Click to view available options
Capacity:
220 g
320 g
420 g
620 g
650 g
820 g
1200 g
2200 g
3200 g
4200 g
5200 g
6200 g
8200 g
Readability:
1 mg
10 mg
100 mg
1000 mg
Model:
CEU Electronic Toploading Balance
Unit Size:
1 unit
Description
- No matter what you're weighing, the new Entris™ II balance is always the right choice.
- Offering unrivalled value and backed by almost 150 years of German engineering expertise, the Entris™ II comes in two product lines, so you can find the balance that meets your specific weighing needs.
- With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
Specifications
Specifications
| Certifications/Compliance | CEU |
| Connection Type | RS232, USB C |
| Capacity | 8200 g |
| Display Type | Touchscreen |
| Readability | 100 mg |
| Material | PBT, Glass, Stainless Steel |
| Model | CEU Electronic Toploading Balance |
| Stabilization Time | ≤1.0 sec. |
| Type | Precision Balance |
| Electrical Requirements | 100 – 240 VAC (±10%), 50 - 60 Hz, 0.2 A |