Learn More
Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification
Alle ansehen Sartorius Produkte
missing translation for 'orderingAttributeHoverText'
Capacity:
60 g
120 g
220 g
320 g
Description:
Verified
missing translation for 'unitSize'
Each
Beschreibung
- Entris™ II Advanced balances - Quality, Value, and Consistency
- For accurate weighing results that can be trusted time after time
- Simplifies balance leveling with built-in real-time level support
- Increases reliability of weighing results using integrated protection systems
- Controlled access to balance settings with user management
- Gapless documentation thanks to CalAuditTrail
Spezifikation
Spezifikation
| Certifications/Compliance | CE Certificate |
| Connection Type | D-sub connector, 9-pin; USB UTL; USB Device |
| Description | Verified |
| Capacity | 60 g |
| Display Type | Intuitive Graphic Touch Technology |
| Readability | 0.1 mg |
| Material | Glass, PBT, Stainless Steel |
| Model | Electronic Analytical Balance |
| Stabilization Time | ≤1.5 sec. |
| Voltage | 100/240 V (±10%) |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur