Learn More
ENTPD5 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26746-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is similar to E-type nucleotidases (NTPases)/ecto-ATPase/apyrases. NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD5 contains 4 apyrase-conserved regions which is characteristic of NTPases.Specifications
| ENTPD5 | |
| Polyclonal | |
| Unconjugated | |
| ENTPD5 | |
| CD39 antigen like 4, CD39L4, ENTPD5, ER UDPase, NTPDase 5, Nucleoside diphosphatase, PCPH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 957 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75356 | |
| ENTPD5 | |
| ENTPD5 Fusion Protein Ag25022 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |