Learn More
Enocyanin, TRC
Product Code. 30371294
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371294 25 g 25g
30382610 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30371294 Supplier Toronto Research Chemicals Supplier No. TRCE26236525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 11029-12-2
Molecular Formula C27H31O16
Molecular Weight (g/mol) 611.53
Specifications
Quantity 25 g
Chemical Name or Material Enocyanin