Learn More
ENO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16421-1-AP-20UL
 View more versions of this product
Product Code. 16806044
  • 1710.33 NOK / 20µL
Description
This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
Specifications
ENO3
Polyclonal
Unconjugated
ENO3
Beta enolase, ENO3, Enolase 3, enolase 3 (beta, muscle), MSE, Muscle specific enolase, Skeletal muscle enolase
Rabbit
Antigen Affinity Chromatography
RUO
13808, 2027, 25438
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P13929, P15429, P21550
ENO3
ENO3 Fusion Protein Ag9613
20 μL
Primary
Human, Mouse, Rat