Learn More
Enmetazobactam, MedChemExpress
Product Code. 16474846
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16474846 100 mg 100mg
16476517 5 mg 5mg
16423798 10 mg 10mg
16420177 50 mg 50mg
4 options
Product Code. 16474846
Brand: Medchem Express HY103095
MedChemExpress Enmetazobactam (AAI101) is an extended-spectrum β-lactamase inhibitor, against many resistant Gram-negative pathogens.
Enmetazobactam (AAI101) is an extended-spectrum β-lactamase inhibitor, against many resistant Gram-negative pathogens.
Specifications
Chemical Name or Material Enmetazobactam
CAS 1001404-83-6
Color White
Purity Grade Notes Research
Molecular Formula C11H14N4O5S
Quantity 100 mg
Synonym AAI101
Solubility Information DMSO : ≥ 113.3 mg/mL (360.46 mM)
SMILES C[N+]1=NN(C[C@@](S([C@]2([H])C3)(=O)=O)(C)[C@H](C([O-])=O)N2C3=O)C=C1
Molecular Weight (g/mol) 314.32
Formula Weight 314.32
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation