Learn More
Macherey-Nagel™ Endotoxin-Free Buffers for NucleoBond™ Xtra EF Kits
Description
The new generation of anion exchanger – ultra-fast, endotoxin-free plasmid DNA with high yield.
- Plasmid DNA with less than 0.05 EU/μg for transfection of highly sensitive cells
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Midi prep in approximately 45 min, Maxi prep in 50 min
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 400 μg (Midi) and 1000 μg (Maxi)
- NucleoBond™ Xtra Plus EF kits contain NucleoBond Finalizers to speed up DNA precipitation
Specifications
Specifications
| Chemical Name or Material | FIL-EF |
| CAS | 64-17-5d |
| Color | Colorless |
| For Use With (Application) | Endotoxin-free plasmid DNA purification |
| For Use With (Equipment) | NucleoBond™ Xtra EF Kit |
| pH | 6.0 to 7.0 |
| Physical Form | Liquid |
| Quantity | 600 mL |
| Formulation | Potassium chloride, Chemicals mixture <1%, Water |
| Odor | Odorless |