Learn More
Lonza™ Clonetics™ Endothelial Cell Growth Medium BulletKit-2-MV
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- EGM™-2MV
- Microvascular Endothelial Cell Growth Medium-2 BulletKit™
Order Info
dimProductType
Specifications
Specifications
| Product Type | Growth Medium |
| Quantity | 1 Kit |
| Content And Storage | No BBE (bovine brain extract), hEGF, hydrocortisone, GA-1000 (Gentamicin, Amphotericin-B), FBS (fetal bovine serum) 25 ml, VEGF, hFGF-B, R3, -IGF-1, ascorbic acid |
Safety and Handling
Product Identifier
- EGM-2 MV Endothel Med BulletKit
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H225-Highly flammable liquid and vapour.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P370+P378-In case of fire: Use to extinguish.
- MIXTURE LIST-Contains : ethanol, gentamycinsulphate