missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Antibody (MEM-226), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-452PE
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Flow (Cell Surface), CyTOF
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| PE | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
| Recombinant full length protein (Human). Expressed in vaccinia virus containing CD105 cDNA. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2022 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunoprecipitation, Flow Cytometry (Cell Surface), CyTOF | |
| MEM-226 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction