Learn More
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, TRC
Product Code. 30482432
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482432 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30482432 Supplier Toronto Research Chemicals Supplier No. TRCM2886001G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R,3r,5S)-9-methyl-9-azabicyclo(3.3.1)nonan-3-amine
CAS 76272-56-5
Molecular Formula C9 H18 N2
Quantity 1 g
Synonym (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]-nonan-3-amine, endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, endo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane, Granisetron Hydrochloride Imp. E (EP)
InChI Formula InChI=1S/C9H18N2/c1-11-8-3-2-4-9(11)6-7(10)5-8/h7-9H,2-6,10H2,1H3/t7-,8+,9-
SMILES CN1[C@@H]2CCC[C@H]1C[C@@H](N)C2
IUPAC Name (1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-amine
Molecular Weight (g/mol) 154.253
Formula Weight 154.147