Learn More
endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, TRC
Product Code. 30479068
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479068 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30479068 Supplier Toronto Research Chemicals Supplier No. TRCM288600100MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (1R,3r,5S)-9-methyl-9-azabicyclo(3.3.1)nonan-3-amine
CAS 76272-56-5
Molecular Formula C9 H18 N2
Quantity 100 mg
Synonym (1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]-nonan-3-amine, endo-9-Methyl-9-azabicyclo[3.3.1]nonan-3-amine, endo-3-Amino-9-methyl-9-azabicyclo[3.3.1]nonane, Granisetron Hydrochloride Imp. E (EP)
InChI Formula InChI=1S/C9H18N2/c1-11-8-3-2-4-9(11)6-7(10)5-8/h7-9H,2-6,10H2,1H3/t7-,8+,9-
SMILES CN1[C@@H]2CCC[C@H]1C[C@@H](N)C2
IUPAC Name (1R,5S)-9-methyl-9-azabicyclo[3.3.1]nonan-3-amine
Molecular Weight (g/mol) 154.253
Formula Weight 154.147