Learn More
SPEX Certiprep™ Extractor for Microvials
Product Code. 15340492 Shop All SPEX Certiprep Products
Change view
Click to view available options
Type:
End Plug Extractor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15340492 End Plug Extractor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15340492 Supplier SPEX Certiprep™ Supplier No. 6758
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use this to easily removes the end plug from frozen microvials.
This end-plug extractor accessory for the SPEX SamplePrep freezer mill 6757 microvial set is a bench tool for easy removal of end plugs from frozen microvials. Simply place three microvials into the extractor cradle and press down on the extractor handle to remove the end plugs. Sample can be retrieved from the vial while it is still cold. Suction cups under the extractor secure it to the benchtop preventing slippage and allowing for one-handed operation.
Specifications
Type End Plug Extractor
Description Holds up to three Microvals
For Use With (Equipment) 6757 Microvial