Learn More
Encenicline hydrochloride, MedChemExpress
Product Code. 16492607
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16492607 50 mg 50mg
16459097 5 mg 5mg
16446766 100 mg 100mg
16426518 10 mg 10mg
4 options
Product Code. 16492607
Brand: Medchem Express HY15430A
MedChemExpress Encenicline hydrochloride (EVP-6124 hydrochloride) is a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors (nAChRs).
Encenicline hydrochloride (EVP-6124 hydrochloride) is a novel partial agonist of α7 neuronal nicotinic acetylcholine receptors (nAChRs).
Specifications
Chemical Name or Material Encenicline hydrochloride
CAS 550999-74-1
Color White
Purity Grade Notes Research
Molecular Formula C16H18Cl2N2OS
Quantity 50 mg
Synonym EVP-6124 hydrochloride
Solubility Information DMSO : ≥ 50 mg/mL (139.94 mM)
SMILES O=C(C1=CC2=C(C(Cl)=CC=C2)S1)N[C@H]3CN4CCC3CC4.[H]Cl
Molecular Weight (g/mol) 357.3
Formula Weight 357.3
Percent Purity 97.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration