Learn More
Day Impex™ Enamelled Steel Container
Product Code. p-8011266
Click to view available options
Capacity (Metric):
200 mL
700 mL
750 mL
1 L
1100 mL
1500 mL
2 L
3 L
4500 mL
10 L
Height (Metric):
215mm
228mm
268mm
275mm
280mm
285mm
290mm
330mm
350mm
445mm
Unit Size:
Each
Product Code. 11817452
Brand: Day Impex™ 200003
Only null left
Add to basket
Add to basket
Safely store and transport liquid gases. Day Impex™ Enamelled Steel Container is specially designed for laboratory, hospital, and industrial use.
  • Safe for use with all liquid gases and ideal for cold traps
  • Attractive blue stove-enamelled steel outer container
Specifications
Diameter (Metric) Outer 68mm
For Use With (Application) Use for liquid gas or as a cold trap
Height (Metric) Inner 170mm
Height (Metric) 215mm
Item Description Without handle or lid
Capacity (Metric) 200 mL
Depth (Metric) Interior 170 mm
Diameter (Metric) Inner 40 mm
Material Enameled Steel