Learn More
Day Impex™ Enamelled Steel Container
Product Code. 11766399
Click to view available options
Capacity (Metric):
200 mL
700 mL
750 mL
1 L
1100 mL
1500 mL
2 L
3 L
4500 mL
10 L
Height (Metric):
215mm
228mm
268mm
275mm
280mm
285mm
290mm
330mm
350mm
445mm
Unit Size:
Each
Product Code. 11766399
Brand: Day Impex™ 200011
Safely store and transport liquid gases. Day Impex™ Enamelled Steel Container is specially designed for laboratory, hospital, and industrial use.
  • Safe for use with all liquid gases and ideal for cold traps
  • Attractive blue stove-enamelled steel outer container
Specifications
Diameter (Metric) Outer 116mm
For Use With (Application) Use for liquid gas or as a cold trap
Height (Metric) Inner 185mm
Height (Metric) 228mm
Includes With lid and handle
Capacity (Metric) 1 L
Depth (Metric) Interior 185 mm
Diameter (Metric) Inner 85 mm
Material Enameled Steel