Learn More
Emetine Dihydrochloride, LoGiCal
Product Code. 30459118
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459118 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30459118 Supplier LoGiCal Supplier No. LGCFOR0131.02
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Emetine Dihydrochloride
CAS 316-42-7
Analyte or Component Names Emetine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C29 H40 N2 O4 . 2 Cl H
Certifications/Compliance ISO 9001
Synonym Emetine Dihydrochloride, (2S,3R,11bS)-3-Ethyl-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-2-[[(1R)-1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolinyl]methyl]-2H-benzo[a]quinolizine hydrochloride (1:2), 3-Ethyl-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-2-[(1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolyl)methyl]-2H-benzo[a]quinolizine dihydrochloride, (2S,3R,11bS)-3-Ethyl-1,3,4,6,7,11b-hexahydro-9,10-dimethoxy-2-[[(1R)-1,2,3,4-tetrahydro-6,7-dimethoxy-1-isoquinolinyl]methyl]-2H-benzo[a]quinolizine dihydrochloride, 6',7',10,11-Tetramethoxy-emetan dihydrochloride, Emetine dihydrochloride, (-)-Emetine dihydrochloride, Emetine hydrochloride, Hemometina, l-Emetine dihydrochloride
InChI Formula InChI=1S/C29H40N2O4.2ClH/c1-6-18-17-31-10-8-20-14-27(33-3)29(35-5)16-23(20)25(31)12-21(18)11-24-22-15-28(34-4)26(32-2)13-19(22)7-9-30-24;;/h13-16,18,21,24-25,30H,6-12,17H2,1-5H3;2*1H/t18-,21-,24+,25-;;/m0../s1
Solution Type Neat
SMILES Cl.Cl.CC[C@H]1CN2CCc3cc(OC)c(OC)cc3[C@@H]2C[C@@H]1C[C@H]4NCCc5cc(OC)c(OC)cc45
IUPAC Name (2S,3R,11bS)-2-[[(1R)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl]methyl]-3-ethyl-9,10-dimethoxy-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizine;dihydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 553.56
Quantity 10 mg
Formula Weight 552.2522 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.