Learn More
Merck Accessories for ZHE Hazardous Waste System
Shop All Merck Products
Click to view available options
Type:
2-Way Valve, 1/8 in. NPTM
Cylinder O-Rings
Luer Fitting
Piston O-Rings, Wiper Seal
Piston with O-Rings, Wiper Seal
Quick-Disconnect Female Swagelok
Quick-Disconnect Male Swagelok
Support Screen 90 mm
Tedlar Bag
Tedlar Bag Adapter
Vent/Relief Valve
ZHE Tight Syringe
Unit Size:
Each
Pack of 10
Pack of 3
Pack of 6
Description
Accessories for filter holders for sample preparation.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | ZHE Hazardous Waste Filtration System |
| Sterility | Non-sterile |
| Max. Pressure | 8.6 |
| Pressure Rating | 8.6 bar (125 psi) |
| Type | Vent/Relief Valve |