Learn More
Merck Accessories for ZHE Hazardous Waste System
Product Code. 10101451 Shop All Merck Products
Change view
Click to view available options
Type:
2-Way Valve, 1/8 in. NPTM
Cylinder O-Rings
Luer Fitting
Piston O-Rings, Wiper Seal
Piston with O-Rings, Wiper Seal
Quick-Disconnect Female Swagelok
Quick-Disconnect Male Swagelok
Support Screen 90 mm
Tedlar Bag
Tedlar Bag Adapter
Vent/Relief Valve
ZHE Tight Syringe
Unit Size:
Each
Pack of 10
Pack of 3
Pack of 6
Product Code. Type unitSize
10101451 Piston with O-Rings, Wiper Seal Each
10768051 2-Way Valve, 1/8 in. NPTM Each
10082143 Cylinder O-Rings Pack of 3
10203451 Piston O-Rings, Wiper Seal Pack of 3
10718441 Luer Fitting Each
10430683 ZHE Tight Syringe Each
10079890 - Pack of 6
10508411 Support Screen 90 mm Each
10708251 Quick-Disconnect Female Swagelok Each
10601872 Tedlar Bag Pack of 10
10602062 Tedlar Bag Adapter Each
10661672 Vent/Relief Valve Each
10718251 Quick-Disconnect Male Swagelok Each
13 options
Product Code. 10101451
Brand: Merck YT3009001
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) ZHE Hazardous Waste filtration system
Material Stainless Steel, Fluoroelastomer
Sterility Non-sterile
Type Piston with O-Rings, Wiper Seal