Learn More
MilliporeSigma™ TLC Sprayer and Accessories
Product Code. 15833881
Change view
Click to view available options
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15833881 Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15833881 Supplier MilliporeSigma™ Supplier No. 1085410001
 Restricted Item
Estimated Shipment: 15-12-2026
TLC accessories allow professionals to work more quickly, more efficiently, and more reliably
Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.
TRUSTED_SUSTAINABILITY
Specifications
Includes 0.8 mm for low viscosity solutions, 1.25 mm optimized for high-viscosity solutions (5 x 0.8 mm bore/1 x 1.25 mm bore)