Learn More
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Accessories for filter holders for sample preparation.
Brand: Merck 5614
381.13 NOK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700Specifications
Not Autoclavable | |
47 mm high Pressure SS filter holder | |
Non-sterile |
Under Drain Screen | |
Stainless Steel (316) |