Learn More
Merck MultiScreen™ Sealing Tape
Shop All Merck Products
Click to view available options
Color:
Clear
Opaque
Unit Size:
Pack of 100
Description
- Opaque sealing tape is used to seal 100μL plate bottoms read in TopCount™ instruments
- Clear sealing tape can be used to seal all manufacturers' 300μL plate tops and plate bottoms for MicroBeta™ readers
- Precut to size
Specifications
Specifications
| Color | Opaque |
| Sterility | Non-sterile |
| No. per Pack | 100 |
| For Use With (Application) | Used to seal 100μL plate bottoms read in TopCount™ instruments |
| Product Type | Sealing Tape |
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion